Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloromethiazide 99.0+%, TCI America™
SDP

Catalog No. T131810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T131810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T131810G Supplier TCI America Supplier No. T131810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133-67-5
Molecular Formula C8H8Cl3N3O4S2
Molecular Weight (g/mol) 380.639
MDL Number MFCD00057315
InChI Key LMJSLTNSBFUCMU-UHFFFAOYSA-N
Synonym trichlormethiazide, trichloromethiazide, naqua, trichlormetazid, metahydrin, achletin, diurese, trichlormethiazid, diu-hydrin, chlopolidine
PubChem CID 5560
ChEBI CHEBI:9683
IUPAC Name 6-chloro-3-(dichloromethyl)-1,1-dioxo-3,4-dihydro-2H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES C1=C2C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)NC(N2)C(Cl)Cl

Specifications

Color White
Quantity 10 g
Formula Weight 380.64
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trichloromethiazide
RTECSNumber DK8925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.