accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 1330-86-5

IUPAC Name: 1,6-bis(6-methylheptyl) hexanedioate
Synonyms: diisooctyl adipate
Molecular Weight (g/mol): 370.57
Molecular Formula: C22H42O4
InChi Key: CJFLBOQMPJCWLR-UHFFFAOYSA-N
SMILES: CC(C)CCCCCOC(=O)CCCCC(=O)OCCCCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results