accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133550-30-8

IUPAC Name: N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Synonyms: N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Molecular Weight (g/mol): 294.31
Molecular Formula: C17H14N2O3
InChi Key: TUCIOBMMDDOEMM-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)NCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results