CAS RN 134052-73-6
IUPAC Name:
4-[amino(carboxy)methyl]benzoic acid
Synonyms:
4-carboxyphenylglycine (S)-4-carboxyphenylglycine 4-cpg 4-carboxylphenylglycine
Molecular Weight (g/mol):
195.17
Molecular Formula:
C9H9NO4
InChi Key:
VTMJKPGFERYGJF-UHFFFAOYNA-N
SMILES:
NC(C(O)=O)C1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
R&D Systems™ (S)-4-Carboxyphenylglycine
CAS: 134052-73-6 Molecular Formula: C9H9NO4