accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNNH(E/Z)(E/Z)

CAS RN 13412-07-2

IUPAC Name: N-[(4-methylphenyl)imino]-N'-(phenylamino)benzenecarboximidamide
Synonyms: N-[(4-methylphenyl)imino]-N'-(phenylamino)benzenecarboximidamide
Molecular Weight (g/mol): 314.39
Molecular Formula: C20H18N4
InChi Key: TYLDKKLAWINVKP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N=NC(=NNC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results