accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NClNOOCH3CH3NOH(E)

CAS RN 134279-78-0

IUPAC Name: (E)-[chloro(1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-imidazol-4-yl)methylidene]amino benzoate
Synonyms: (E)-[chloro(1-hydroxy-2,2,5,5-tetramethylimidazol-4-yl)methylidene]amino benzoate
Molecular Weight (g/mol): 323.78
Molecular Formula: C15H18ClN3O3
InChi Key: JBKQCCCCMAUIRG-LDADJPATSA-N
SMILES: CC1(C)N=C(\C(Cl)=N/OC(=O)C2=CC=CC=C2)C(C)(C)N1O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)