accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3CNONOHNNH3C

CAS RN 134296-40-5

IUPAC Name: hydrogen N-{8-methyl-8-azabicyclo[3.2.1]octan-3-yl}-2-oxo-3-(propan-2-yl)-2,3-dihydro-1H-1,3-benzodiazole-1-carboxamide chloride
Synonyms: hydrogen 3-isopropyl-N-{8-methyl-8-azabicyclo[3.2.1]octan-3-yl}-2-oxo-1,3-benzodiazole-1-carboxamide chloride
Molecular Weight (g/mol): 378.9
Molecular Formula: C19H27ClN4O2
InChi Key: NQYXXIUVFVOJCX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)N1C(=O)N(C(=O)NC2CC3CCC(C2)N3C)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results