accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1343364-70-4

IUPAC Name: ({[({[3,4-dihydroxy-5-(4-oxo-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid tetrasodium
Synonyms: ({[3,4-dihydroxy-5-(4-oxo-2-sulfanylidene-3H-pyrimidin-1-yl)oxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxyphosphonic acid tetrasodium
Molecular Weight (g/mol): 592.16
Molecular Formula: C9H15N2Na4O14P3S
InChi Key: ZMGCYKHWZHBMKG-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].[Na].OC1C(O)C(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N1C=CC(=O)NC1=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results