Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2-ThioUTP tetrasodium salt
SDP

Catalog No. 32801 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-801 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-801 Supplier Tocris Bioscience™ Supplier No. 3280/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective P2Y2 agonist

Potent and selective P2Y2 agonist (EC50 values are 0.035, 0.35 and 1.5 μM for hP2Y2, hP2Y4 and hP2Y6 receptors respectively).

Chemical Identifiers

CAS 1343364-70-4
Molecular Formula C9H15N2Na4O14P3S

Specifications

Quantity 1 mg
Formula Weight 588.13
Purity 95%
Molecular Weight (g/mol) 592.159
Chemical Name or Material 2-ThioUTP tetrasodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.