accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHSiCH3CH3H3C

CAS RN 13435-12-6

IUPAC Name: N-(trimethylsilyl)acetamide
Synonyms: acetamide, N-(trimethylsilyl)-
Molecular Weight (g/mol): 131.25
Molecular Formula: C5H13NOSi
InChi Key: LWFWUJCJKPUZLV-UHFFFAOYSA-N
SMILES: CC(=O)N[Si](C)(C)C
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)