Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Trimethylsilylacetamide 98.0+%, TCI America™
SDP

Catalog No. T059025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T059025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T059025G Supplier TCI America Supplier No. T059025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13435-12-6
Molecular Formula C5H13NOSi
Molecular Weight (g/mol) 131.25
MDL Number MFCD00008671
InChI Key LWFWUJCJKPUZLV-UHFFFAOYSA-N
Synonym n-trimethylsilyl acetamide, trimethylsilylacetamide, acetamide, n-trimethylsilyl, tms acetamide, acetylamino trimethylsilane, trimethylsilyl acetamide, acetamide, tms derivative, n-trimethylsilyl-acetamide, n-tms-acetamide, n-trimethylsilylacetoamide
PubChem CID 25989
IUPAC Name N-trimethylsilylacetamide
SMILES CC(=O)N[Si](C)(C)C

Specifications

Color White
Quantity 25 g
UN Number 1325
Formula Weight 131.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Trimethylsilylacetamide
RTECSNumber AD0250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.