accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaAlOSiOOOSiOO

CAS RN 1344-00-9

IUPAC Name: aluminium(3+) sodium bis(oxosilanebis(olate))
Synonyms: sodium aluminosilicate sodium silicoaluminate
Molecular Weight (g/mol): 202.14
Molecular Formula: AlNaO6Si2
InChi Key: URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)