accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13446-53-2

IUPAC Name: dibromomagnesium hexahydrate
Synonyms: magnesium bromide hexahydrate
Molecular Weight (g/mol): 292.2
Molecular Formula: Br2H12MgO6
InChi Key: LGLXXNHIGIJYQQ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.Br[Mg]Br
Molecular Weight (g/mol) 
  • (2)
  • (9)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Grade 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (9)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)