Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bromide hexahydrate, 99%, extra pure

Catalog No. AC446252500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446252500 250 g Plastic bottle
AC446250010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446252500 Supplier Thermo Scientific Chemicals Supplier No. 446252500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-53-2
Molecular Formula Br2H12MgO6
Molecular Weight (g/mol) 292.20
MDL Number MFCD00149780
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
PubChem CID 17749097
SMILES O.O.O.O.O.O.Br[Mg]Br

Specifications

Melting Point 165.0°C
Quantity 250 g
Linear Formula MgBr2·6H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
IUPAC Name magnesium;dibromide;hexahydrate
Formula Weight 292.21
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium bromide hexahydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.