accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13472-84-9

IUPAC Name: 3-chloro-2-methoxypyridine
Synonyms: 3-chloro-2-methoxypyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: MSICGKUDCSUMJZ-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results