Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methoxypyridine 97.0+%, TCI America™
SDP

Catalog No. C25655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25655G 5 g
C25651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25655G Supplier TCI America Supplier No. C25655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13472-84-9
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03095227
InChI Key MSICGKUDCSUMJZ-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxy-pyridine, pyridine, 3-chloro-2-methoxy, 2-methoxy-3-chloropyridine, pubchem1189, acmc-1c0pw, ksc494g7h, 3-chloranyl-2-methoxy-pyridine
PubChem CID 13295749
IUPAC Name 3-chloro-2-methoxypyridine
SMILES COC1=C(C=CC=N1)Cl

Specifications

Color Yellow
Boiling Point 53°C
Quantity 5 g
UN Number 1993
Formula Weight 143.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.