accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13473-01-3

IUPAC Name: 5-chloro-2-methoxypyridine
Synonyms: 5-chloro-2-methoxypyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: NPYYXUYLIHZYOU-UHFFFAOYSA-N
SMILES: COC1=NC=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results