Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. C25225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25225G 5 g
C25221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25225G Supplier TCI America Supplier No. C25225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13473-01-3
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD06254388
InChI Key NPYYXUYLIHZYOU-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxy-pyridine, pyridine, 5-chloro-2-methoxy, 2-methoxy-5-chloro pyridine, pubchem6602, 2-methoxy-5-chloropyridine, acmc-1c02r, ksc494g7j, 5-chloranyl-2-methoxy-pyridine, 5-chloro-2-methoxypyridine
PubChem CID 4738276
IUPAC Name 5-chloro-2-methoxypyridine
SMILES COC1=NC=C(C=C1)Cl

Specifications

Color Yellow
Boiling Point 180°C
Quantity 5 g
UN Number 1993
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.