accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOOHOO

CAS RN 13504-85-3

IUPAC Name: 1-[(benzyloxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 265.27
Molecular Formula: C13H15NO5
InChi Key: WWVCWLBEARZMAH-UHFFFAOYNA-N
SMILES: OC1CC(N(C1)C(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)