accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135062-02-1

IUPAC Name: 2-ethoxy-4-({[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]carbamoyl}methyl)benzoic acid
Synonyms: repaglinide prandin novo nordisk brand 2 of repaglinide repaglinida repaglinide preparation (+-)-repaglinide novo nordisk brand of repaglinide NovoNorm repaglinide (substance) repaglinide (product)
Molecular Weight (g/mol): 452.6
Molecular Formula: C27H36N2O4
InChi Key: FAEKWTJYAYMJKF-QHCPKHFHSA-N
SMILES: CCOC1=CC(CC(=O)N[C@@H](CC(C)C)C2=CC=CC=C2N2CCCCC2)=CC=C1C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results