Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Repaglinide

Catalog No. AC463230010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463230010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463230010 Supplier Thermo Scientific Chemicals Supplier No. 463230010

missing translation for 'validMainframeMsg'

CAS: 135062-02-1 | C27H36N2O4 | 452.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135062-02-1
Molecular Formula C27H36N2O4
Molecular Weight (g/mol) 452.60
MDL Number MFCD00906179
InChI Key FAEKWTJYAYMJKF-QHCPKHFHSA-N
IUPAC Name 2-ethoxy-4-({[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]carbamoyl}methyl)benzoic acid
SMILES CCOC1=CC(CC(=O)N[C@@H](CC(C)C)C2=CC=CC=C2N2CCCCC2)=CC=C1C(O)=O

Specifications

Melting Point 129.0°C to 130.0°C
Quantity 1 g
Formula Weight 452.59
Chemical Name or Material Repaglinide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.