CAS RN 1351761-44-8
IUPAC Name:
N4-ethyl-N2-[5-fluoro-2-methoxy-4-(morpholine-4-carbonyl)phenyl]-5-(trifluoromethyl)pyrimidine-2,4-diamine
Synonyms:
N4-ethyl-N2-[5-fluoro-2-methoxy-4-(morpholine-4-carbonyl)phenyl]-5-(trifluoromethyl)pyrimidine-2,4-diamine
Molecular Weight (g/mol):
443.4
Molecular Formula:
C19H21F4N5O3
InChi Key:
XCFLWTZSJYBCPF-UHFFFAOYSA-N
SMILES:
CCNC1=NC(NC2=C(OC)C=C(C(=O)N3CCOCC3)C(F)=C2)=NC=C1C(F)(F)F