CAS RN 1351813-81-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefminox Sodium Heptahydrate, G-Biosciences
CAS: 1351813-81-4 Molecular Weight (g/mol): 667.66 g/mol