CAS RN 1352827-50-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ N-methylated pomalidomide
Functionalized cereblon ligand for generating Degrader negative controls; also used as Pomalidomide negative control