accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OF

CAS RN 135427-08-6

IUPAC Name: 4-fluoro-3-methylbenzaldehyde
Synonyms: 4-fluoro-3-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: NRFKZFFVTGGEQF-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results