accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3O

CAS RN 13562-76-0

IUPAC Name: butan-2-yl 3-oxobutanoate
Synonyms: sec-butyl 3-oxobutanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: QSTNBMLCULGCQE-UHFFFAOYNA-N
SMILES: CCC(C)OC(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results