accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135634-18-3

NHNOHFFHCl(E/Z)

CAS RN 135634-18-3

IUPAC Name: N-[(2,4-difluorophenyl)(piperidin-4-yl)methylidene]hydroxylamine hydrochloride
Synonyms: N-[(2,4-difluorophenyl)(piperidin-4-yl)methylidene]hydroxylamine hydrochloride
Molecular Weight (g/mol): 276.71
Molecular Formula: C12H15ClF2N2O
InChi Key: CPVWKXXFKMUDPA-UHFFFAOYSA-N
SMILES: Cl.ON=C(C1CCNCC1)C1=C(F)C=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results