accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFF

CAS RN 135734-59-7

IUPAC Name: 4-ethyl-4'-[4-(trifluoromethoxy)phenyl]-1,1'-bi(cyclohexane)
Synonyms: 4-ethyl-4'-[4-(trifluoromethoxy)phenyl]-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 354.46
Molecular Formula: C21H29F3O
InChi Key: PBIFTROFCZATFC-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(CC1)C1=CC=C(OC(F)(F)F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results