accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONNN

CAS RN 13578-51-3

IUPAC Name: 1-(4-methylbenzenesulfonyl)-1H-1,2,4-triazole
Synonyms: 1-(4-methylbenzenesulfonyl)-1,2,4-triazole
Molecular Weight (g/mol): 223.25
Molecular Formula: C9H9N3O2S
InChi Key: GFWABQNSSIQCLB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=NC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results