accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135884-31-0

IUPAC Name: {1-[(tert-butoxy)carbonyl]-1H-pyrrol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)pyrrol-2-ylboronic acid
Molecular Weight (g/mol): 211.02
Molecular Formula: C9H14BNO4
InChi Key: ZWGMJLNXIVRFRJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)