Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-2-pyrroleboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B40635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4063-5G 5 g
B40631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4063-5G Supplier TCI America Supplier No. B40635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135884-31-0
Molecular Formula C9H14BNO4
MDL Number MFCD01318939
InChI Key ZWGMJLNXIVRFRJ-UHFFFAOYSA-N
PubChem CID 2734321

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Synonym n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid, 1-boc-pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1 h-pyrrol-2-ylboronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid
SMILES CC(C)(C)OC(=O)N1C=CC=C1B(O)O
IUPAC Name {1-[(tert-butoxy)carbonyl]-1H-pyrrol-2-yl}boronic acid
Molecular Weight (g/mol) 211.02
Formula Weight 211.02
Physical Form Crystalline Powder
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.