accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNFFFFFFOHFN

CAS RN 1359164-11-6

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-[2-fluoro-4'-({4-[(pyridin-4-yl)methyl]piperazin-1-yl}methyl)-[1,1'-biphenyl]-4-yl]propan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-(2-fluoro-4'-{[4-(pyridin-4-ylmethyl)piperazin-1-yl]methyl}-[1,1'-biphenyl]-4-yl)propan-2-ol
Molecular Weight (g/mol): 527.49
Molecular Formula: C26H24F7N3O
InChi Key: KVHKWAZUPPBMLL-UHFFFAOYSA-N
SMILES: OC(C1=CC(F)=C(C=C1)C1=CC=C(CN2CCN(CC3=CC=NC=C3)CC2)C=C1)(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)