accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13596-46-8

IUPAC Name: λ²-cobalt(2+) diammonium disulfate
Synonyms: λ²-cobalt(2+) diammonium disulfate
Molecular Weight (g/mol): 287.12
Molecular Formula: CoH8N2O8S2
InChi Key: NUHXWPRATKOXDB-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].[Co++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results