accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136112-00-0

IUPAC Name: 3-methylbutan-1-ol; phenol; trichloromethane
Synonyms: chloroform; isoamyl alcohol; phenol
Molecular Weight (g/mol): 301.63
Molecular Formula: C12H19Cl3O2
InChi Key: ZYWFEOZQIUMEGL-UHFFFAOYSA-N
SMILES: ClC(Cl)Cl.CC(C)CCO.OC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
pH 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)