CAS RN 136194-77-9
IUPAC Name:
3-{23-methyl-14-oxo-3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-3-yl}propanenitrile
Synonyms:
3-{23-methyl-14-oxo-3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-3-yl}propanenitrile
Molecular Weight (g/mol):
378.44
Molecular Formula:
C24H18N4O
InChi Key:
VWVYILCFSYNJHF-UHFFFAOYSA-N
SMILES:
CN1C2=C(C=CC=C2)C2=C1C1=C(C3=C2C(=O)NC3)C2=CC=CC=C2N1CCC#N
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ InSolution™ Gö 6976
Inhibitor of protein kinase C (PKC; IC50 = 7
2
MilliporeSigma™ Calbiochem™ Gö 6976
Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC50 = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC50 = 2.3 nM) and βI (IC50 = 6.2 nM).