accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOOSSOHCH3Br

CAS RN 136310-93-5

IUPAC Name: 7-{[2-hydroxy-2,2-bis(thiophen-2-yl)acetyl]oxy}-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide
Synonyms: tiotropium bromide
Molecular Weight (g/mol): 472.41
Molecular Formula: C19H22BrNO4S2
InChi Key: DQHNAVOVODVIMG-UHFFFAOYNA-M
SMILES: [Br-].C[N+]1(C)C2CC(CC1C1OC21)OC(=O)C(O)(C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results