accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13679-73-7

IUPAC Name: 1-(4-methylthiophen-2-yl)ethan-1-one
Synonyms: 1-(4-methylthiophen-2-yl)ethanone
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: NFZNHUNPIJPRMP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results