accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13683-41-5

IUPAC Name: (1-bromoethenyl)trimethylsilane
Synonyms: (1-bromoethenyl)trimethylsilane
Molecular Weight (g/mol): 179.13
Molecular Formula: C5H11BrSi
InChi Key: VVDJVCJVVHHCIB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(Br)=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)