CAS RN 13707-88-5
IUPAC Name:
hydrogen 1-[2-(prop-2-en-1-yl)phenoxy]-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms:
alprenolol hydrochloride alprenolol hcl
Molecular Weight (g/mol):
285.81
Molecular Formula:
C15H24ClNO2
InChi Key:
RRCPAXJDDNWJBI-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C)NCC(O)COC1=CC=CC=C1CC=C
1
–
1
of
1
results
1
R&D Systems™ Alprenolol hydrochloride
CAS: 13707-88-5 Molecular Formula: C15H24ClNO2