accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1371569-69-5

IUPAC Name: 4-[3-(benzyloxy)phenyl]-2-(ethanesulfinyl)-6-(trifluoromethyl)pyrimidine
Synonyms: 4-[3-(benzyloxy)phenyl]-2-(ethanesulfinyl)-6-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 406.42
Molecular Formula: C20H17F3N2O2S
InChi Key: NTDFYGSSDDMNHI-UHFFFAOYNA-N
SMILES: CCS(=O)C1=NC(=CC(=N1)C(F)(F)F)C1=CC(OCC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results