CAS RN 13718-22-4
CAS RN 13718-22-4
IUPAC Name:
dirubidium(1+) tetraoxomolybdenumdiuide
Synonyms:
dirubidium(1+) tetraoxomolybdenumdiuide
Molecular Weight (g/mol):
330.88
Molecular Formula:
MoO4Rb2
InChi Key:
JTWQMTQQNPXYGJ-UHFFFAOYSA-N
SMILES:
[Rb+].[Rb+].O=[Mo--](=O)(=O)=O
1
–
1
of
1
results
1
Rubidium molybdenum oxide, 99.5% (Metals basis), Thermo Scientific™
CAS: 13718-22-4 Molecular Formula: Rb2MoO4 MDL Number: MFCD00049518 Synonym: Rubidium molybdate