accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNONH2OOCH3

CAS RN 13726-52-8

IUPAC Name: 1,5-diethyl 2-[(4-aminophenyl)formamido]pentanedioate
Synonyms: 1,5-diethyl 2-[(4-aminophenyl)formamido]pentanedioate
Molecular Weight (g/mol): 322.36
Molecular Formula: C16H22N2O5
InChi Key: RJXFBLRRPYBPTM-UHFFFAOYNA-N
SMILES: CCOC(=O)CCC(NC(=O)C1=CC=C(N)C=C1)C(=O)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results