accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137281-23-3

IUPAC Name: (2R)-2-{[4-(2-{2-amino-4-oxo-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)phenyl]formamido}pentanedioic acid
Synonyms: pemetrexed LY231514 disodium rolazar tifolar pemetrexed disodium
Molecular Weight (g/mol): 427.42
Molecular Formula: C20H21N5O6
InChi Key: WBXPDJSOTKVWSJ-CYBMUJFWSA-N
SMILES: NC1=NC(=O)C2=C(NC=C2CCC2=CC=C(C=C2)C(=O)N[C@H](CCC(O)=O)C(O)=O)N1
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results