accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COOHF

CAS RN 1373168-89-8

IUPAC Name: 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Synonyms: 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Weight (g/mol): 266.08
Molecular Formula: C13H16BFO4
InChi Key: WVKOYVWFDCAHKF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C(=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results