CAS RN 137339-65-2
CAS RN 137339-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Thrombin Receptor Agonist
The Thrombin Receptor Agonist, also referenced under CAS 137339-65-2, controls the biological activity of Thrombin Receptor. This small molecule/inhibitor is primarily used for Activators/Inducers applications.