accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHReOOO

CAS RN 13768-11-1

IUPAC Name: rheniumoylol
Synonyms: perrhenic acid
Molecular Weight (g/mol): 251.21
Molecular Formula: HO4Re
InChi Key: UGSFIVDHFJJCBJ-UHFFFAOYSA-M
SMILES: O[Re](=O)(=O)=O
Molecular Weight (g/mol) 
  • (14)
  • (2)
Grade 
  • (2)
  • (6)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results