Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Rhenium (Re) Standard for AAS 1000 ppm in 5% HNO3, Ricca Chemical
SDP

Catalog No. RV010146500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010146500 500 mL
RV010146100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010146500 Supplier Ricca Chemical Company Supplier No. RV010146500N

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Chemical Identifiers

CAS 13768-11-1 , 7697-37-2 , 7732-18-5
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
PubChem CID 83718
IUPAC Name rheniumoylol
SMILES O[Re](=O)(=O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 94.87, 5, 0.13
CAS Max % 94.87, 5, 0.13
Density 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Chemical Name or Material VeriSpec™ Rhenium (Re) Standard for AAS 1000 ppm in 5% HNO3, Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
ShelfLife 24 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.