accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOVOO

CAS RN 13769-43-2

IUPAC Name: potassium vanadiumoylolate
Synonyms: potassium dioxidovanadium(1+)
Molecular Weight (g/mol): 138.04
Molecular Formula: KO3V
InChi Key: BQFYGYJPBUKISI-UHFFFAOYSA-N
SMILES: [K+].[O-][V](=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)