CAS RN 137813-35-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cyclo(Arg-Gly-Asp-D-Phe-Val)
Potent inhibitor of cell adhesion.