CAS RN 138-52-3
D-(-)-Salicin, 99%
CAS: 138-52-3 Molecular Formula: C13H18O7 Molecular Weight (g/mol): 286.28 MDL Number: MFCD00006590 InChI Key: NGFMICBWJRZIBI-UJPOAAIJSA-N Synonym: salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside PubChem CID: 439503 ChEBI: CHEBI:17814 IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol SMILES: C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
Salicin, MP Biomedicals
CAS: 138-52-3 Molecular Formula: C13H18O7 Molecular Weight (g/mol): 286.28 MDL Number: MFCD00006590 InChI Key: NGFMICBWJRZIBI-UJPOAAIJSA-N Synonym: salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside PubChem CID: 439503 ChEBI: CHEBI:17814 IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol SMILES: C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
D(-)-Salicin, 99+%
CAS: 138-52-3 Molecular Formula: C13H18O7 Molecular Weight (g/mol): 286.27 MDL Number: MFCD00006590 InChI Key: NGFMICBWJRZIBI-UJPOAAIJSA-N Synonym: salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside PubChem CID: 439503 ChEBI: CHEBI:17814 IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol SMILES: C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
phyproof™ Salicin, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Salicin 98.0+%, TCI America™
CAS: 138-52-3 Molecular Formula: C13H18O7 Molecular Weight (g/mol): 286.28 MDL Number: MFCD00006590 InChI Key: NGFMICBWJRZIBI-UJPOAAIJSA-N Synonym: salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside PubChem CID: 439503 ChEBI: CHEBI:17814 IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol SMILES: C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O